DNA transfection reagent, PolyMag Neo™
Supplier: OZ BIOSCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PG60100 PG60200 PG61000
OZBIPG60100EA 132 GBP
OZBIPG60100 OZBIPG60200 OZBIPG61000
DNA transfection reagent, PolyMag Neo™
Transfection Reagents
PolyMag Neo™ transfection reagent is specifically designed to achieve high transfection efficiency combined with superior transgene expression level. It is composed of magnetic nanoparticles coated with cationic molecules. It is the ideal transfection reagent for a wide variety of cells.
  • High transgene expression
  • High transfection efficiency
  • Multipurpose (various types of nucleic acids)
  • Universal (primary cells and cell lines)
  • Simple, ready to use and rapid
  • Non toxic
  • Compatible with and without serum-containing culture media
Store at +4 °C.
Accessories information: This reagent needs to be used with a magnetic plate.
Delivery information: Shipped at room temperature.