Freund's incomplete adjuvant, IFAVax

Supplier: OZ BIOSCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

IFA0010 IFA0050 IFA0100
OZBIIFA0010EA 45.3 GBP
OZBIIFA0010 OZBIIFA0050 OZBIIFA0100
Freund's incomplete adjuvant, IFAVax
Protein Expression Protein Expression Reagents

Incomplete Freund’s Adjuvant (IFA) is a water-in-oil emulsion that is similar to the complete Freund’s Adjuvant (CFA) without addition of heat-killed mycobacteria (Mycobacterum butyricum).


  • Suitable for the stimulation of Th2 response and antibody production


It consists of a mixture of mineral oil and emulsifier in a ratio of 85% v/v oil and 15% v/v emulsifier. It is not a pre-formed emulsion and thus it must be mixed with an equal volume of aqueous solution of antigen and subsequently emulsified prior to use. The emulsifier is mannide monooleate, an ester consisting of mannitol as the hydrophilic residue and oleic acid (unsaturated C18 fatty acid) as the hydrophobic part.


Store at +4 °C


Delivery information: Shipped at room temperature.


Caution: Intended for research use only.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR