Bradford protein assay kits (B-Pak)
Supplier: OZ BIOSCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BA00050 BA00070 BA00100
OZBIBA00050EA 127 GBP
OZBIBA00050 OZBIBA00070 OZBIBA00100
Bradford protein assay kits (B-Pak)
Assays Protein Assays
Bradford Protein Assay Kit (B-Pak) is a straightforward and rapid kit for determining the concentration of proteins in solution. This ready to use kit is provided with 1X reagent and 2 sets of BSA standards which means no dilution, filtration or calculation required. B-Pak is provided with adapted protocols for all experimental conditions – micro and macroassays.
  • Accurate determination of protein concentration
  • Ready to use prediluted standard protein
  • Cheaper than competitors
  • Convenient packaging
  • Fast and easy to use
Store at +4 °C.
Delivery information: Shipped at room temperature.