Ag-AK, Immunofluorescence test, IMAGEN™
Supplier: THERMO FISHER DIAGNOSTICS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S610830-2 S610930-2 S611030-2 S611130-2 S611230-2 S611330-2 S611430-6 S612130-2
OXOIS610830-2EA 85 GBP
OXOIS610830-2 OXOIS610930-2 OXOIS611030-2 OXOIS611130-2 OXOIS611230-2 OXOIS611330-2 OXOIS611430-6 OXOIS612130-2
Ag-AK, Immunofluorescence test, IMAGEN™
Microbiology Test Systems Antigen-Antibody Tests Ag-AK, Immunoassay
IMAGEN™ is a qualitative direct immunofluorescence test used for detection and confirmation of the presence of virus in cell culture or in clinical specimen.
  • Very specific