Quanti-Cult Plus™ Salmonella enterica subsp. enterica serovar Typhimurium ATCC™ 14028™
Supplier: THERMO FISHER DIAGNOSTICS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
R4716000
OXOIR4716000EA 1270 GBP
OXOIR4716000
Quanti-Cult Plus™ Salmonella enterica subsp. enterica serovar Typhimurium ATCC™ 14028™
Microbiology Test Systems Microorganism Tests
Quanti-Cult Plus™ Salmonella enterica subsp. enterica serovar Typhimurium ATCC™ 14028™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
Packaging: Includes 10 preserved organism vials and 10 rehydration vials.
Order Now
SPECIFICATIONS
Environmentally Preferable