Culti-Loops™ Escherichia coli ATCC™ 11775™
Supplier: THERMO FISHER DIAGNOSTICS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
R4601989
OXOIR4601989EA 231 GBP
OXOIR4601989
Culti-Loops™ Escherichia coli ATCC™ 11775™
Microbiology Test Systems Microorganism Tests
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Order Now
SPECIFICATIONS
Environmentally Preferable