Culti-Loops™ Paenibacillus macerans ATCC™ 8509™
Supplier: THERMO FISHER DIAGNOSTICS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
R4601237
OXOIR4601237EA 231 GBP
OXOIR4601237
Culti-Loops™ Paenibacillus macerans ATCC™ 8509™
Microbiology Test Systems Microorganism Tests
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.