Culti-Loops™ Staphylococcus aureus ATCC™ BAA-1026™
Supplier: THERMO FISHER DIAGNOSTICS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
R4601026
OXOIR4601026EA 231 GBP
OXOIR4601026
Culti-Loops™ Staphylococcus aureus ATCC™ BAA-1026™
Microbiology Test Systems Microorganism Tests
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.