McFarland turbidity standards, Remel™
Supplier: THERMO FISHER DIAGNOSTICS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
R20411 R20412 R20413 R20414 R20421 R20410
OXOIR20411EA 82.1 GBP
OXOIR20411 OXOIR20412 OXOIR20413 OXOIR20414 OXOIR20421 OXOIR20410
McFarland turbidity standards, Remel™
Standards Turbidity Standards
These McFarland equivalence turbidity standard is packed in plastic case with a visual comparision card. Tube size of 15×103 mm.
  • Fits in most spectrometers
  • Polystyrene technology allows longer shelf life
Delivery information: The turbidity standard set includes 1 each of 0,5, 1,0, 2,0, 3,0 and 4,0.