Wafer tweezers, perforated
Supplier: OUTILS RUBIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
39S-4
OUTI39S-4EA 70.9 GBP
OUTI39S-4
Wafer tweezers, perforated
Forceps
Stainless steel. These tweezers are specially designed to handle different fragile and delicate materials of gallium or silicon wafers.
  • Perforated lower paddle allows liquids to drop off
  • Smooth polished non glare surface on the inside of the paddel in order to prevent chipping and marring the wafer
  • Satin finish on the outside of the paddle for glare prevention