Anti-CD20 IgG2a Monoclonal Antibody [clone: 743X65]

Supplier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA591088
ORIGTA591088EA 628 GBP
ORIGTA591088
Anti-CD20 IgG2a Monoclonal Antibody [clone: 743X65]
Antibodies
Recommended Dilutions: Use at 0.1-1ug/ml. Flow Cytometry , Direct cell killing

Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 743X65
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD20
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 743X65
Reactivity Human
Host Mouse
Isotype IgG2a
Flow cytometry Yes
Immunogen A full length cDNA gene delivered by DNA immunization
Form Concentrated
Purification Protein A purification
Storage buffer PBS with 50% glycerol and 0,05% azide
Storage temperature –20 °C
Shipping temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR