Anti-SAT2 Mouse Monoclonal Antibody [clone: OTI1C9]

Supplier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA504633
ORIGTA504633EA 583 GBP
ORIGTA504633
Anti-SAT2 Mouse Monoclonal Antibody [clone: OTI1C9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SAT2 (spermidine/spermine N1-acetyltransferase family member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR