Anti-TET3 Mouse Monoclonal Antibody [clone: 3B3]

Supplier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CF803928
ORIGCF803928EA 781 GBP
ORIGCF803928
Anti-TET3 Mouse Monoclonal Antibody [clone: 3B3]
Antibodies
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: TET3
Clonality: Monoclonal
Clone: 3B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR