Anti-CTDSP1 Mouse Monoclonal Antibody [clone: 1G2]

Supplier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CF502227
ORIGCF502227EA 781 GBP
ORIGCF502227
Anti-CTDSP1 Mouse Monoclonal Antibody [clone: 1G2]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: CTDSP1
Clonality: Monoclonal
Clone: 1G2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR