Anti-APC (Ab-1) (FE9) Mouse Monoclonal Antibody

Supplier: Merck Millipore (Oncogene)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OP44-100
OP44-100EA 497 GBP
OP44-100
Anti-APC (Ab-1) (FE9) Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: APC (Ab-1) (FE9)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR