Anti-PMAIP1 Mouse Monoclonal Antibody [clone: 114C307]

Supplier: Merck Millipore (Oncogene)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OP180-100
OP180-100EA 522 GBP
OP180-100
Anti-PMAIP1 Mouse Monoclonal Antibody [clone: 114C307]
Antibodies
Type: Primary
Antigen: PMAIP1 (phorbol-12-myristate-13-acetate-induced protein 1)
Clonality: monoclonal
Clone: 114C307
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR