Anti-c-ErbB2/c-Neu (Ab-4) (7.16.4) Mouse Monoclonal Antibody [clone: 7.16.4]

Supplier: Merck Millipore (Oncogene)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OP16L-100
OP16L-100EA 465 GBP
OP16L-100
Anti-c-ErbB2/c-Neu (Ab-4) (7.16.4) Mouse Monoclonal Antibody [clone: 7.16.4]
Antibodies
Type: Primary
Antigen: c-ErbB2/c-Neu (Ab-4) (7.16.4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR