cfDNA DNA kit, CE IVD, Mag-Bind®

Supplier: OMEGA BIO-TEK
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M3298-01CEIVD M3298-02CEIVD
OMEGM3298-01CEIVDEA 728 GBP
OMEGM3298-01CEIVD OMEGM3298-02CEIVD
cfDNA DNA kit, CE IVD, Mag-Bind®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

The Mag-Bind® cfDNA Kit CE IVD is designed for rapid and reliable isolation of circulating cell-free DNA from 1 to 4 ml plasma/serum samples.


The Mag-Bind® cfDNA Kit CE IVD is designed for rapid and reliable isolation of circulating cell-free DNA from 1 to 4 ml plasma/serum samples. The Mag-Bind® cfDNA Kit CE IVD can be processed manually with 15 ml centrifuge tubes or on automated platforms with appropriate plasticware. The procedure eliminates the need for funnels and vacuum steps providing hands-free operation in automated protocols. The uniquely formulated binding buffer from Omega Bio-Tek allows for large sample volumes to be processed in automated formats with 4 ml serum or plasma being processed in a 24-well plate. The magnetic properties of the Mag-Bind® Particles CH enable fast magnetic separation, especially during steps involving large volumes. The high-binding capability decreases the amount of magnetic particles required thereby reducing the elution volume i.e., cfDNA from up to 4 ml serum or plasma can be eluted in just 50 μl.


Certifications: CE-IVD IVDR

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR