Upright microscopes, CX41
Supplier: OLYMPUS OPTICAL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
K23008948 K23008950 K23008951
OLYMK23008948EA 0 GBP
OLYMK23008948 OLYMK23008950 OLYMK23008951
Upright microscopes, CX41
Microscopes
These microscopes set the standards in both optics and design.
  • UIS2 plan achromatic objectives
  • Camera can be adapted to trinocular models
  • High image clarity: Brightfield, phase contrast, fluorescence
Designed for applications in biology and medicine.
Ordering information: Supplied with a protective cover and two spare bulbs.
Order Now
SPECIFICATIONS
Environmentally Preferable
Nosepiece Quintuple
Condenser Abbe N.A. 1,25
Illumination Halogen