COURIER™ 3000 Shipping Scales
Supplier: OHAUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30745885 30746021 30745886 30745887 30745888 30745889 30746022 30746023 30746025 30809911 30809912 30809913 30809914 30809915
OHAU30745885EA 170 GBP
OHAU30745885 OHAU30746021 OHAU30745886 OHAU30745887 OHAU30745888 OHAU30745889 OHAU30746022 OHAU30746023 OHAU30746025 OHAU30809911 OHAU30809912 OHAU30809913 OHAU30809914 OHAU30809915
COURIER™ 3000 Shipping Scales
Balances and Scales
The Courier 3000 Series shipping scales intuitive, user-friendly interfaces, bright displays, and low-profile steel construction ensure durability and reliability from offices to warehouses to even veterinary offices (the Courier 3000 is effective for weighing small animals). Versatile and effective.
  • For Weighing and display hold
  • 4-digit, 7-segment LCD display with white backlight, 28 mm high digits
  • ABS indicator, powder-coated steel platform and frame
  • 150% overload protection
  • AC adapter included
  • 100 Hours with disposable batteries
  • Painted steel platform construction