Multifunctional column mounted washdown bench scales, for standard industrial applications, Defender 5000
Supplier: OHAUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30444826 30444827 30471149 30444819 30471146 30444822 30444829 30471150 30444830 30461436 30461437 30461568 30461569 30461570 30461571 30461572 30461573 30461574 30461575 30461576 30461577 30461578 30471144 30471145 30471147 30471148 30444824 30444821 30444823 30444828 30444818 30444820 30471151 30471158 30471159 30471160 30471161 30471162 30471163 30471164 30471165 30444825
OHAU30444826EA 1850 GBP
OHAU30444826 OHAU30444827 OHAU30471149 OHAU30444819 OHAU30471146 OHAU30444822 OHAU30444829 OHAU30471150 OHAU30444830 OHAU30461436 OHAU30461437 OHAU30461568 OHAU30461569 OHAU30461570 OHAU30461571 OHAU30461572 OHAU30461573 OHAU30461574 OHAU30461575 OHAU30461576 OHAU30461577 OHAU30461578 OHAU30471144 OHAU30471145 OHAU30471147 OHAU30471148 OHAU30444824 OHAU30444821 OHAU30444823 OHAU30444828 OHAU30444818 OHAU30444820 OHAU30471151 OHAU30471158 OHAU30471159 OHAU30471160 OHAU30471161 OHAU30471162 OHAU30471163 OHAU30471164 OHAU30471165 OHAU30444825
Multifunctional column mounted washdown bench scales, for standard industrial applications, Defender 5000
Balances and Scales
The Defender 5000 series multifunctional bench scales are ideal for numerous applications including production, packaging, inventory and shipping. Durably constructed to withstand harsh environments and equipped with a stainless steel platform, stainless steel frame and a stainless steel (single range models) or aluminium load cell (dual range models).
  • Large backlit display and alpha-numeric keypad with function keys
  • Indicator guides user with on-screen prompts
  • Multiple application modes and high storage capacity for greater flexibility
  • Multiple
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable