Digital block heaters, Advanced

Supplier: OHAUS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

30392082 30392061 30392089 30392096
OHAU30392082EA 825 GBP
OHAU30392082 OHAU30392061 OHAU30392089 OHAU30392096
Digital block heaters, Advanced
Block Heaters Digital Block Heaters
Designed for applications that require repeatable results and superior temperature stability. These multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a wide variety of other laboratory procedures.

  • PID temperature control, with optional external RTD probe, offers a temperature stability as low as ±0,1 °C with a temperature uniformity as low as ±0,1 °C
  • Hot warning symbol light is illuminated when the temperature is above 40 °C
  • Overshoot protection ensures unit will
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR