Heavy duty orbital shakers, digital
Supplier: OHAUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30391811 30391844 30391865 30391872 30391879 30391886
OHAU30391811EA 2290 GBP
OHAU30391811 OHAU30391844 OHAU30391865 OHAU30391872 OHAU30391879 OHAU30391886
Heavy duty orbital shakers, digital
Shakers Orbital Shakers
These digital heavy duty orbital shakers are designed for a wide range of applications, including cell cultures, that require accurate and repeatable results. The microprocessor control provides consistent uniform shaking while safely ramping to the set speed. Ideal for cell cultures, solubility studies, and extraction procedures. The large capacity shakers have a more powerful drive mechanism and larger orbits for optimal shaking of large vessels.
  • Exceptional speed control, accuracy and durability
  • RS232 interface provides two-way communication for data
More Product Information