Syringe filters, Target2™
Supplier: THERMO NATIONAL SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F2500-3 F2504-3 F2513-3 F2513-4 F2513-1 F2500-1 F2500-12 F2500-13 F2500-14 F2500-15 F2500-16 F2500-17 F2500-2 F2500-4 F2500-5 F2500-50 F2500-6 F2500-7 F2500-8 F2502-1 F2502-2 F2502-3 F2504-1 F2504-15 F2504-16 F2504-2 F2504-4 F2504-5 F2504-6 F2504-7 F2504-8 F2513-14 F2513-15 F2513-16 F2513-17 F2513-2 F2513-5 F2513-6 F2513-7 F2513-8
NSCAF2500-3EA 453 GBP
NSCAF2500-3 NSCAF2504-3 NSCAF2513-3 NSCAF2513-4 NSCAF2513-1 NSCAF2500-1 NSCAF2500-12 NSCAF2500-13 NSCAF2500-14 NSCAF2500-15 NSCAF2500-16 NSCAF2500-17 NSCAF2500-2 NSCAF2500-4 NSCAF2500-5 NSCAF2500-50 NSCAF2500-6 NSCAF2500-7 NSCAF2500-8 NSCAF2502-1 NSCAF2502-2 NSCAF2502-3 NSCAF2504-1 NSCAF2504-15 NSCAF2504-16 NSCAF2504-2 NSCAF2504-4 NSCAF2504-5 NSCAF2504-6 NSCAF2504-7 NSCAF2504-8 NSCAF2513-14 NSCAF2513-15 NSCAF2513-16 NSCAF2513-17 NSCAF2513-2 NSCAF2513-5 NSCAF2513-6 NSCAF2513-7 NSCAF2513-8
Syringe filters, Target2™
Filters Syringe Filters
These syringe filters have a PP housing.
  • Each lot is tested for both physical properties and UV detectable extractables
  • Solvent resistant
  • Membranes are available with 1,0 μm glass prefilter for high solid samples