Screw thread vial convenience kits
Supplier: THERMO NATIONAL SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C4015-88
NSCAC4015-88EA 209 GBP
NSCAC4015-88
Screw thread vial convenience kits
Vials
These convenience vial kits contain caps with pre-inserted septa that are not attached to vials.
  • Dust-free
  • Unassembled vial kit
Ordering information: These kits include 100 vials and 100 caps with pre-assembled septa.
Packaging: Packaged in polybags.