Anti-SPINK1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP2-29620 NBP2-29620SS
NOVUNBP2-29620EA 427 GBP
NOVUNBP2-29620 NOVUNBP2-29620SS
Anti-SPINK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SPINK1 (serine peptidase inhibitor, Kazal type 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR