Anti-TLR6 Mouse Monoclonal Antibody (Biotin) [clone: 86B1153.2]

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP2-24791
NOVUNBP2-24791EA 693 GBP
NOVUNBP2-24791
Anti-TLR6 Mouse Monoclonal Antibody (Biotin) [clone: 86B1153.2]
Antibodies
Type: Primary
Antigen: TLR6 (toll-like receptor 6)
Clonality: monoclonal
Clone: 86B1153.2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR