Anti-ERManI Mouse Monoclonal Antibody (DyLight® 550) [clone: 3C2]

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP2-13167R
NOVUNBP2-13167REA 583 GBP
NOVUNBP2-13167R
Anti-ERManI Mouse Monoclonal Antibody (DyLight® 550) [clone: 3C2]
Antibodies
Type: Primary
Antigen: ERManI
Clonality: monoclonal
Clone: 3C2
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR