Anti-CHORDC1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP1-78304 NBP1-78304SS
NOVUNBP1-78304EA 539 GBP
NOVUNBP1-78304 NOVUNBP1-78304SS
Anti-CHORDC1 Rabbit Polyclonal Antibody
Antibodies
The CHORDC1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CHORDC1. This antibody reacts with human, mouse. The CHORDC1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:150

Type: Primary
Antigen: CHORDC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR