Anti-RUNX2 Rabbit Polyclonal Antibody (DyLight® 550)

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP1-77461R
NOVUNBP1-77461REA 703 GBP
NOVUNBP1-77461R
Anti-RUNX2 Rabbit Polyclonal Antibody (DyLight® 550)
Antibodies
Type: Primary
Antigen: RUNX2 (runt-related transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR