Anti-Ferroportin 1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP1-21502B
NOVUNBP1-21502BEA 785 GBP
NOVUNBP1-21502B
Anti-Ferroportin 1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: Ferroportin 1
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

SPECIFICATIONS

Antigen symbol FPN1
Environmentally Preferable
Antigen name Ferroportin 1
Antigen synonyms HFE4|SLC11A3|IREG1|SLC40A1|MST079|solute carrier family 40 member 1|MSTP079|MTP1|FPN1
Antibody type Primary
Clonality Polyclonal
Conjugation Biotin
Reactivity Human
Host Rabbit
Gene ID 30061
Isotype
Western blot Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR