Anti-Oval Cell Marker Rat Monoclonal Antibody (DyLight® 650) [clone: OC2-1D11]

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP1-18963C
NOVUNBP1-18963CEA 650 GBP
NOVUNBP1-18963C
Anti-Oval Cell Marker Rat Monoclonal Antibody (DyLight® 650) [clone: OC2-1D11]
Antibodies
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-1D11
Conjugation: DyLight® 650
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR