Anti-HPi3 Mouse Monoclonal Antibody (DyLight® 350) [clone: HIC1-7H10]

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NBP1-18947UV
NOVUNBP1-18947UVEA 650 GBP
NOVUNBP1-18947UV
Anti-HPi3 Mouse Monoclonal Antibody (DyLight® 350) [clone: HIC1-7H10]
Antibodies
Type: Primary
Antigen: HPi3
Clonality: monoclonal
Clone: HIC1-7H10
Conjugation: DyLight® 350
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR