Anti-SAT1 Rabbit Polyclonal Antibody (DyLight® 405)

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB110-41622V
NOVUNB110-41622VEA 704 GBP
NOVUNB110-41622V
Anti-SAT1 Rabbit Polyclonal Antibody (DyLight® 405)
Antibodies
Type: Primary
Antigen: SAT1 (spermidine/spermine N1-acetyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR