Anti-PLIN3 Rabbit Polyclonal Antibody (DyLight® 650)

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB110-40764C
NOVUNB110-40764CEA 703 GBP
NOVUNB110-40764C
Anti-PLIN3 Rabbit Polyclonal Antibody (DyLight® 650)
Antibodies
Type: Primary
Antigen: PLIN3 (perilipin 3)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR