Anti-RNF216 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB100-56891
NOVUNB100-56891EA 423 GBP
NOVUNB100-56891
Anti-RNF216 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RNF216 (ring finger protein 216)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR