Anti-LOX Rabbit Polyclonal Antibody (FITC (Fluorescein))

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB100-2527F
NOVUNB100-2527FEA 748 GBP
NOVUNB100-2527F
Anti-LOX Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antibodies
Type: Primary
Antigen: LOX (lysyl oxidase)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR