9-Fluorenylmethyl chloroformate (Fmoc chloride)

Supplier: Novabiochem (Part of Merck)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

8.52259.0025 8.52259.0100
NOVA8.52259.0025EA 70.1 GBP
NOVA8.52259.0025 NOVA8.52259.0100
9-Fluorenylmethyl chloroformate (Fmoc chloride)
9-Fluorenylmethyl chloroformate (Fmoc chloride)
Formula: C₁₅H₁₁ClO₂
MDL Number: MFCD00001138
CAS Number: 28920-43-6
EINECS: 249-313-6
UN: 1759
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR