Cleanroom wipes, BioClean IsoPure Plus™
Supplier: Nitritex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BIWP-9
NITRBIWP-9EA 617 GBP
NITRBIWP-9
Cleanroom wipes, BioClean IsoPure Plus™
Wipes Cleanroom Wipes Dry Cleanroom Wipes
This wipe is made of double-knit continuous-filament polyester. It features excellent absorption properties, durability and strength.
  • Ultrasonically sealed borders to prevent fibre release
  • Smooth and durable
Suitable for use in Class 10, ISO 4, M2.5 cleanrooms.
Colour: White
Data accurate at time of publishing, latest test results and test methods can be found on supplier's website.
Caution: Data accurate at time of publishing, latest test results and test methods can be found on supplier's website.