Dishes and lids
Supplier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
605 640 644
NICK605EA 28.2 GBP
NICK605 NICK640 NICK644
Dishes and lids
Trays/Dishes/Pots Laboratory Trays/Dishes
Made of aluminium.
  • Very good temperature resistance
  • Good chemical resistance
  • Lightweight