Scoops
Supplier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4020 4021 4022 4023
231-0359EA 111 GBP
231-0359 NICK4021 NICK4022 NICK4023
Scoops
Scoops
Stainless steel. Round bottom. Ideal for use as weighing boats etc.
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Smooth surfaces for fast, easy cleaning