PIG® fibreglass chemical trays
Supplier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PAK739-WH PAK741-WH PAK742-WH PAK743-WH
NEWPPAK739-WHEA 36.3 GBP
NEWPPAK739-WH NEWPPAK741-WH NEWPPAK742-WH NEWPPAK743-WH
PIG® fibreglass chemical trays
Trays/Dishes/Pots Spillage Trays/Pans
Catch drips and control messes with these smooth, seamless fibreglass trays.
  • Reinforced thermoset composite fibreglass resists chemicals and retains its shape, even under heavy loads
  • Smooth corners and seamless design allows trays to be steam cleaned or sterilized for repeated use
  • Molded edges safely contain minor spills or leaks from containers or oily parts
Chemical-resistant trays are easy to clean.