Drum spill trays, PIG®
Supplier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
DRM369 DRM371
NEWPDRM369EA 139 GBP
NEWPDRM369 NEWPDRM371
Drum spill trays, PIG®
Trays/Dishes/Pots Spillage Trays/Pans
HDPE trays contain messy drips and spills from 210 litre drums.
  • Strong, lightweight polyethylene won't rust or corrode
  • Octagonal shape saves space between drums
  • 210 litre drum in place, trays holds 19 litres
Catch drips, leaks and spills that can make your floor a big mess.