Ultra-Low temperature freezers, CryoCube F101h
Supplier: NEW BRUNSWICK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F101341002 F101342002 F101343002 F101344002 F101345002
NEWBF101341002EA 0 GBP
NEWBF101341002 NEWBF101342002 NEWBF101343002 NEWBF101344002 NEWBF101345002
Ultra-Low temperature freezers, CryoCube F101h
Freezers Ultra Low Temperature Freezers
The CryoCube F101h ULT freezers offers a personal storage solution in a convenient package. The compact design means the unit will fit below a standard laboratory bench so samples are always securely close-at-hand. F101h has 2 inner compartments for storing for up to 6 racks/60 boxes giving space for up to 6000 vessels (1,5 ml). These freezers with efficient and reliable 2-stage compressor systems help reduce energy consumption, which helps to lower costs. Energy consumption: (–70/–80 °C): 4,6/6,3 KWh/day.
  • Green cooling liquids (R170/R290) with low Global Warming Potential (GWP)
  • Easy-
More Product Information