Automated cell counter, ADAM-CellT
Supplier: NANOENTEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ADAM-CELLT-UK
NANOADAM-CELLT-UKEA 19710 GBP
NANOADAM-CELLT-UK
Automated cell counter, ADAM-CellT
Cell Counters and Flow Cytometers Automated Cell Counters
ADAM-CellT is an automated cell counter for cGMP. It takes only 25 seconds for ADAM-CellT to present results. ADAM-CellT has an easy-to-use built-in tablet and complies with 21 CFR part 11. When 21 CFR part 11 mode is activated, data cannot be modified by any user.
  • Automated image analysis
  • Accurate result
  • Sensitve CMOS detection and precise automatic stage
  • Auto focusing
  • Cell therapy quality control
To be used with 4 channel Accuchip slides.
Certifications: Compliance with 21 CFR part 11.