Quality control test suspensions
Supplier: CROSSTEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
GP25-03 GP25-07 GP25-08 GP25-09 GP25-11
NAMSGP25-03EA 135 GBP
NAMSGP25-03 NAMSGP25-07 NAMSGP25-08 NAMSGP25-09 NAMSGP25-11
Quality control test suspensions
Microbiology Test Systems Microorganism Tests
The control suspension (growth promotion) contains <100 CFU`s/0,1 ml.
  • 2,5 ml pro vial
  • 25 dose