Biological indicators, spore coupons
Supplier: CROSSTEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CPN-06 CPS-06
NAMSCPN-06EA 694 GBP
NAMSCPN-06 NAMSCPS-06
Biological indicators, spore coupons
Sterilisation Indicators
Spore coupons for use in monitoring plasma/vapourised hydrogen peroxide (VH₂O₂) sterilisation processes.
These biological indicators are designed for monitoring processes whether placed directly in to the chamber, positioned in areas of a device which are the most difficult to sterilise or distributed throughout rooms and isolators.
Spore coupons are supplied with a hole to allow them to hang in the steriliser or room when removed from packaging. Coupons have a rigid structure which allows for easy insertion and removal from a device.
Coupon dimension: 34×7×0,9 mm.
Certifications:More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable