Bottles, wide neck, square, lab quality, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
312110-0008
NALG312110-0008EA 897 GBP
NALG312110-0008
Bottles, wide neck, square, lab quality, with screw cap, Nalgene®
Bottles Plastic Bottles
Wide mouth bottles made of PPCO with uniform walls which is resistant to splitting or puncturing.
  • Good chemical and temperature resistance
  • Wide mouth for easy filling and pouring
  • Leakproof, linerless polypropylene caps
Bottles are autoclavable.
Certifications: Compliance with ISO 13485:2003; 21CFR177.1520 and USP Class VI.
Delivery information: Bottles are supplied with PP linerless closures.