Tanks, cylindrical, with stopcock and cover, Nalgene®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
11102-0005 11102-0007 11102-0010 11102-0015 11102-0030 11102-0055
NALG11102-0005EA 206 GBP
NALG11102-0005 NALG11102-0007 NALG11102-0010 NALG11102-0015 NALG11102-0030 NALG11102-0055
Tanks, cylindrical, with stopcock and cover, Nalgene®
Tanks Cylindrical Tanks
These heavy-duty cylindrical LLDPE tanks with cover and spigot can withstand most acid, alcohols and bases. Tanks used at elevated temperatures or with high specific-gravity liquids should be externally supported.
  • Economical
  • Impact- and abrasion-resistant
  • Not autoclavable