Storage and Presentation Boxes, with Hinged Lid, V5
Supplier: MULTIROIR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
41305 41309 41321 41326 41332 41335 41340 41344 41392BOCBV592
MULC41305EA 2.4 GBP
MULC41305 MULC41309 MULC41321 MULC41326 MULC41332 MULC41335 MULC41340 MULC41344 MULC41392BOCBV592
Storage and Presentation Boxes, with Hinged Lid, V5
Boxes Storage Boxes
Manufactured in crystal polystyrene, these boxes feature an excellent transparent clarity and a high quality finish.
All models are available in standard black base and crystal lid, but are also available with a crystal base and lid.
  • Ideal for the presentation of products
  • Transparent lid and black background for great visibility
  • Available in different sizes